Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allylchlorodimethylsilane 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: AllyldimethylchlorosilaneADMCS

B22237-09 B22237-14 B22237-09 B22237-06 B22237-06 B22237-14
AAAB22237-09EA 130.83 USD InStock
CAAAB22237-14 AAAB22237-09 AAB22237-14 AAB22237-06 CAAAAB22237-09 CAAAB22237-06
Allylchlorodimethylsilane 95%
Allylchlorodimethylsilane


Formula: C₅H₁₁ClSi
Boiling Pt: 110…112 °C
Density: 0.896
Flash Pt: 5 °C(41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000506
CAS Number: 4028-23-3
EINECS: 223-706-2
UN: 2985
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty