Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Pentafluorophenyl)acetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pentafluorobenzyl cyanide(2,3,4,5,6-Pentafluorophenyl)acetonitrile2,3,4,5,6-Pentafluorobenzyl cyanide2-(Perfluorophenyl)acetonitrile

B22239-03 B22239-03 B22239-06 B22239-06
AAB22239-03EA 28.5 USD InStock
CAAAB22239-06 CAAAB22239-03 AAB22239-03 AAB22239-06
(Pentafluorophenyl)acetonitrile 98%
(Pentafluorophenyl)acetonitrile


Formula: (C₆F₅)CH₂CN
MW: 207.1 g/mol
Boiling Pt: 110…111 °C/17 mm
Density: 1.610
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013814
CAS Number: 653-30-5
EINECS: 211-498-6
UN: 3276
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty