Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BAPTA tetrasodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt hydrateTetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate hydrateBAPTA-Na4 hydrate

B22258-06 B22258-14 B22258-03 B22258-03 B22258-06 B22258-14
AAB22258-06EA 635.99 USD InStock
CAAAB22258-14 CAAAB22258-03 AAB22258-06 AAB22258-03 AAB22258-14 CAAAB22258-06
BAPTA tetrasodium salt hydrate 98%
BAPTA tetrasodium salt hydrate
BAPTA tetrasodium salt hydrate is a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research).

BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.


Formula: C₂₂H₂₀N₂Na₄O₁₀.xH₂O
MW: 564.37 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD02683087
CAS Number: 126824-24-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty