Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-4-(Trifluoromethyl)mandelic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-4-(Trifluoromethyl)phenylglycolic acid

B22264-03 B22264-06 B22264-03 B22264-06
AAB22264-03EA 32.97 USD InStock
CAAAB22264-06 AAB22264-03 CAAAB22264-03 AAB22264-06
(±)-4-(Trifluoromethyl)mandelic acid 98%
(±)-4-(Trifluoromethyl)mandelic acid


Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00009724
CAS Number: 395-35-7
EINECS: 206-900-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty