Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Aspartic acid monosodium salt monohydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium L-aspartate monohydrateL-Aspartic acid sodium salt monohydrate

B22321-36 B22321-22 B22321-36 B22321-22
AAB22321-22EA 53.09 USD InStock
CAAAB22321-36 AAB22321-22 AAB22321-36 CAAAB22321-22
L-Aspartic acid monosodium salt monohydrate 99%
L-Aspartic acid monosodium salt monohydrate


Formula: C₄H₆NNaO₄.H₂O
MW: 173.11 g/mol (155.10 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00152960
CAS Number: 323194-76-9
EINECS: 223-264-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty