Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dihexyl sulfide 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Di-n-hexylsulphideDi-n-hexylsulfide

B22325-22 B22325-14 B22325-14 B22325-22
AAB22325-14EA 58.09 USD InStock
AAB22325-14 AAB22325-22 CAAAB22325-14 CAAAB22325-22
Dihexyl sulfide 97%
Dihexyl sulphide


Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 260 °C
Melting Pt: –28.3 °C
Density: 0.849
Flash Pt: 116 °C(241 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009528
CAS Number: 6294-31-1
EINECS: 228-556-1
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty