Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenicarbazide 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Phenylsemicarbazide

B22331-22 B22331-14 B22331-22 B22331-14
AAB22331-22EA 155.99 USD InStock
CAAAB22331-22 CAAAB22331-14 AAB22331-22 AAB22331-14
Phenicarbazide 99%
Phenicarbazide


Formula: C₇H₉N₃O
MW: 151.17 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00007942
CAS Number: 103-03-7
EINECS: 203-072-3
Merck Index: 12,07378
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty