Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-4-aminobutyric Acid4-(tert-Butoxycarbonylamino)butyric Acid4-(Boc-amino)butyric AcidBoc-GABA-OH

B22407-06 B22407-06 B22407-03 B22407-03
AAB22407-06EA 149.99 USD InStock
CAAAB22407-06 AAB22407-06 CAAAB22407-03 AAB22407-03
N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid


Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00037313
CAS Number: 57294-38-9
EINECS: 628-652-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty