Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxy-4-(methacryloyloxy)benzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

B22445-22 B22445-22 B22445-06 B22445-14 B22445-06 B22445-14
AAB22445-14EA 339.99 USD InStock
AAB22445-14 AAB22445-22 CAAAB22445-06 CAAAB22445-22 CAAAB22445-14 AAB22445-06
2-Hydroxy-4-(methacryloyloxy)benzophenone 99%
2-Hydroxy-4-(methacryloyloxy)benzophenone


Formula: C₁₇H₁₄O₄
Melting Pt: 77…79 °C
Density: 1.18
Storage Temperature: Ambient
MDL Number: MFCD00080561
CAS Number: 2035-72-5
EINECS: 218-000-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty