Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Aminosalicylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Amino-2-hydroxybenzoic acid

B22478-02 B22478-77 B22478-02 B22478-77
AAB22478-77EA 79.39 USD InStock
AAB22478-77 AAB22478-02 CAAAB22478-77 CAAAB22478-02
3-Aminosalicylic acid 97%
3-Aminosalicylic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010299
CAS Number: 570-23-0
EINECS: 209-328-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty