Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cholecalciferol (Vitamin D3) 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Vitamin D3Colecalciferol

B22524-06 B22524-06 B22524-03 B22524-03
AAB22524-06EA 270.49 USD InStock
CAAAB22524-03 AAB22524-06 AAB22524-03 CAAAB22524-06
Cholecalciferol (Vitamin D3) 99%
Cholecalciferol (Vitamin D3)


Formula: C₂₇H₄₄O
MW: 384.65 g/mol
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00078131
CAS Number: 67-97-0
EINECS: 200-673-2
UN: 2811
ADR: "6.1",III
Merck Index: 14,10019
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty