Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl methoxyacetate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methoxyacetic methyl ester

B22528-22 B22528-36 B22528-22 B22528-36
AAB22528-22EA 83.39 USD InStock
CAAAB22528-36 CAAAB22528-22 AAB22528-22 AAB22528-36
Methyl methoxyacetate 99%
Methyl methoxyacetate


Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 129…130 °C
Density: 1.051
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008451
CAS Number: 6290-49-9
EINECS: 228-539-9
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty