Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Methylthio)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Methylthio)acetic acid(Methylthio)ethansäure

B22531-06 B22531-14 B22531-03 B22531-06 B22531-14 B22531-03
AAB22531-06EA 97.19 USD InStock
AAB22531-06 CAAAB22531-14 CAAAB22531-03 AAB22531-14 AAB22531-03 CAAAB22531-06
(Methylthio)acetic acid 98%
(Methylthio)acetic acid


Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 99…100 °C/4 mm
Melting Pt: 13…14 °C
Density: 1.224
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075444
CAS Number: 2444-37-3
EINECS: 219-483-6
UN: 3334
ADR: "9",
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty