Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylthiazolidine 98%

Supplier: Thermo Scientific Chemicals
Warning

B22537-14 B22537-14
AAB22537-14EA 222.99 USD InStock
CAAAB22537-14 AAB22537-14
2-Methylthiazolidine 98%
2-Methylthiazolidine


Formula: C₄H₉NS
MW: 103.18 g/mol
Boiling Pt: 76…78 °C/36 mm
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053110
CAS Number: 24050-16-6
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty