Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(4-Chlorobenzoyl)pyridine 99+%

Supplier: Thermo Scientific Chemicals

B22551-03 B22551-06 B22551-03 B22551-06
AAB22551-03EA 67.99 USD InStock
CAAAB22551-06 AAB22551-03 AAB22551-06 CAAAB22551-03
4-(4-Chlorobenzoyl)pyridine 99+%
4-(4-Chlorobenzoyl)pyridine


Formula: C₁₂H₈ClNO
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006431
CAS Number: 14548-48-2
EINECS: 238-587-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty