Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-N-methylaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

B22565-09 B22565-30 B22565-18 B22565-18 B22565-09 B22565-30
AAB22565-18EA 309.99 USD InStock
CAAAB22565-09 AAB22565-18 CAAAB22565-18 AAB22565-09 AAB22565-30 CAAAB22565-30
3-Fluoro-N-methylaniline 97%
3-Fluoro-N-methylaniline


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 94 °C/19 mm
Density: 1.11
Flash Pt: 86 °C(187 °F)
Storage Temperature: Ambient
MDL Number: MFCD02683095
CAS Number: 1978-37-6
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty