Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-tert-Butyl-L-tyrosine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Tyr(tBu)-OH

B22575-03 B22575-06 B22575-03 B22575-06
AAB22575-03EA 77.09 USD InStock
CAAAB22575-06 AAB22575-03 CAAAB22575-03 AAB22575-06
O-tert-Butyl-L-tyrosine 99%
O-tert-Butyl-L-tyrosine


Formula: C₁₃H₁₉NO₃
MW: 237.30 g/mol
Melting Pt: ca 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037202
CAS Number: 18822-59-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty