Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methoxy-2-methylbenzothiazole 97%

Supplier: Thermo Scientific Chemicals

B22584-06 B22584-03 B22584-06 B22584-03
AAB22584-06EA 118.99 USD InStock
CAAAB22584-03 CAAAB22584-06 AAB22584-06 AAB22584-03
5-Methoxy-2-methylbenzothiazole 97%
5-Methoxy-2-methylbenzothiazole


Formula: C₉H₉NOS
Melting Pt: 34…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00082664
CAS Number: 2941-69-7
EINECS: 220-929-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty