Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Heptylbenzoic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

B22589-06 B22589-14 B22589-14 B22589-06
AAB22589-06EA 66.09 USD InStock
AAB22589-06 AAB22589-14 CAAAB22589-06 CAAAB22589-14
4-Heptylbenzoic acid 99+%
4-Heptylbenzoic acid


Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00009722
CAS Number: 38350-87-7
EINECS: 253-894-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty