Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyloctanoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-methylcaprylic acid

B22616-06 B22616-14 B22616-14 B22616-06
AAB22616-06EA 84.59 USD InStock
AAB22616-06 AAB22616-14 CAAAB22616-06 CAAAB22616-14
4-Methyloctanoic acid 98%
4-Methylcaprylic acid


Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 272 °C
Density: 0.910
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051938
CAS Number: 54947-74-9
EINECS: 259-404-2
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty