Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercaptothiophene 97% (as monomer) contains dimer

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Thiophene-2-thiol

B22642-06 B22642-03 B22642-14 B22642-06 B22642-03 B22642-14
AAB22642-06EA 105.99 USD InStock
AAB22642-06 CAAAAB22642-03 AAB22642-14 CAAAB22642-14 CAAAB22642-06 AAAB22642-03
2-Mercaptothiophene 97% (as monomer) contains dimer
2-Mercaptothiophene


Formula: C₄H₄S₂
MW: 116.20 g/mol
Boiling Pt: 62…65 °C/12 mm
Density: 1.247
Flash Pt: 66 °C(151 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051666
CAS Number: 7774-74-5
EINECS: 231-881-1
UN: 3334
ADR: 9,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty