Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Amino-1,2-propanediol 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-3-Amino-1,2-dihydroxy propane

B22672-22 B22672-14 B22672-36 B22672-14 B22672-36 B22672-22
AAB22672-14EA 67.89 USD InStock
AAB22672-14 AAB22672-36 CAAAB22672-14 CAAAB22672-36 CAAAB22672-22 AAB22672-22
(±)-3-Amino-1,2-propanediol 97%
(±)-3-Amino-1,2-propanediol


Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 264…266 °C/739 mm
Density: 1.186
Flash Pt: 186 °C(366 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty