Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-o-toluidine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Amino-6-fluorotoluene 3-Fluoro-2-methylaniline

B22764-06 B22764-14 B22764-14 B22764-06
AAB22764-14EA 214.99 USD InStock
CAAAB22764-14 AAB22764-14 AAB22764-06 CAAAB22764-06
3-Fluoro-o-toluidine 98+%
3-Fluoro-o-toluidine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 205 °C
Melting Pt: 7 °C
Density: 1.099
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007760
CAS Number: 443-86-7
EINECS: 207-142-4
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty