Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Bromosuccinic acid 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Bromo-1,4-butanedioic acid

B22829-06 B22829-06 B22829-14 B22829-22 B22829-22 B22829-14
AAB22829-22EA 231.99 USD InStock
CAAAB22829-06 AAB22829-22 CAAAB22829-14 AAB22829-14 CAAAB22829-22 AAB22829-06
(±)-Bromosuccinic acid 96%
(±)-Bromosuccinic acid


Formula: HOOCBrCHCH₂COOH
MW: 196.99 g/mol
Melting Pt: 164…166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004213
CAS Number: 923-06-8
EINECS: 213-087-7
Merck Index: 12,01461
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty