Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citronellic acid 94%

Supplier: Thermo Scientific Chemicals
Warning

B22862-14 B22862-14 B22862-36 B22862-36 B22862-22 B22862-22
AAB22862-14EA 87.28 USD InStock
CAAAB22862-14 AAB22862-14 CAAAB22862-36 AAB22862-36 AAB22862-22 CAAAB22862-22
Citronellic acid 94%
Citronellic acid


Formula: C₁₀H₁₈O₂
Boiling Pt: 121…122 °C/1 mm
Density: 0.923
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002728
CAS Number: 502-47-6
EINECS: 207-939-7
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty