Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Tetrahydro-2-furoic acid 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Tetrahydrofuran Carboxylic AcidTetrahydrofuran-2-carboxylic acid

B22927-14 B22927-22 B22927-22 B22927-14
AAB22927-14EA 63.69 USD InStock
CAAAB22927-22 CAAAB22927-14 AAB22927-14 AAB22927-22
(±)-Tetrahydro-2-furoic acid 97+%
(±)-Tetrahydro-2-furoic acid


Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 244 °C
Density: 1.213
Flash Pt: 139 °C(282 °F)
Storage Temperature: Ambient
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty