Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl ethyl ether 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Ethoxy-2-methylpropane

B22945-22 B22945-06 B22945-22 B22945-14 B22945-06 B22945-14
AAB22945-22EA 629 USD InStock
CAAAB22945-06 AAB22945-22 CAAAB22945-14 CAAAB22945-22 AAB22945-06 AAB22945-14
tert-Butyl ethyl ether 99%
tert-Butyl ethyl ether


Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 72…73 °C
Melting Pt: –94 °C
Density: 0.750
Flash Pt: -19 °C(-2 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009225
CAS Number: 637-92-3
EINECS: 211-309-7
UN: 1179
ADR: "3",II
Merck Index: 13,03809
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty