Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Acetyl-γ-butyrolactone 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Acetyl-γ-butyrolactone3-Acetyldihydrofuran-2(3H)-one2-Acetylbutyrolactone

B22967-22 B22967-22 B22967-36 B22967-36
AAB22967-22EA 32.39 USD InStock
CAAAB22967-36 AAB22967-22 AAB22967-36 CAAAB22967-22
α-Acetyl-γ-butyrolactone 98+%
α-Acetyl-γ-butyrolactone


Formula: C₆H₈O₃
MW: 128.13 g/mol
Boiling Pt: 107…108 °C/5 mm
Density: 1.190
Flash Pt: 135 °C(275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005394
CAS Number: 517-23-7
EINECS: 208-235-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty