Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Di-p-toluoyl-L-tartaric acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric AcidDi-p-toluoyl-L-tartaric acid(-)-O,O'-Di-p-toluoyl-L-tartaric acid

B22972-22 B22972-14 B22972-14 B22972-06 B22972-22
AAB22972-06EA 48.14 USD InStock
AAB22972-06 CAAAB22972-14 AAB22972-22 CAAAB22972-22 AAB22972-14
(-)-Di-p-toluoyl-L-tartaric acid 98%
(-)-Di-p-toluoyl-L-tartaric acid


Formula: C₂₀H₁₈O₈
MW: 386.36 g/mol
Melting Pt: 166…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00064440
CAS Number: 32634-66-5
EINECS: 251-131-7
Order Now
Specification Test Results
Optical rotation -139° (c=1 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty