Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc D-gluconate hydrate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Gluconic acid Zzinc(II) salt hydrateZinc(II) gluconate hydrateZinc gluconate hydrate

B23022-22 B23022-36
AAB23022-22EA 56.59 USD InStock
AAB23022-22 AAB23022-36
Zinc D-gluconate hydrate 97%
Zinc D-gluconate hydrate


Formula: C₁₂H₂₂O₁₄Zn·xH₂O
MW: 455.69 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00868110
CAS Number: 4468-02-4
EINECS: 224-736-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty