Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexaphenylcyclotrisiloxane 98+%

Supplier: Thermo Scientific Chemicals

B23061-14 B23061-14 B23061-06 B23061-06
AAB23061-14EA 296.99 USD InStock
AAB23061-14 AAB23061-06 CAAAB23061-14 CAAAB23061-06
Hexaphenylcyclotrisiloxane 98+%
Hexaphenylcyclotrisiloxane


Formula: C₃₆H₃₀O₃Si₃
Boiling Pt: 300 °C/1 mm
Melting Pt: ca 175…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00023110
CAS Number: 512-63-0
EINECS: 208-145-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty