Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Thiopheneboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Thienylboronic acidThiophene-2-boronic acidThiophen-2-ylboronic acid

B23071-03 B23071-06 B23071-14 B23071-14 B23071-06 B23071-03
AAB23071-14EA 313.99 USD InStock
AAB23071-14 AAB23071-03 AAB23071-06 CAAAB23071-03 CAAAB23071-14 CAAAB23071-06
2-Thiopheneboronic acid 98%
2-Thiopheneboronic acid


Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00151850
CAS Number: 6165-68-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty