Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethynylcyclopentanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

B23109-06 B23109-22 B23109-14 B23109-22 B23109-14 B23109-06
AAB23109-22EA 656.43 USD InStock
AAB23109-22 CAAAB23109-06 AAB23109-14 AAB23109-06 CAAAB23109-14 CAAAB23109-22
1-Ethynylcyclopentanol 98+%
1-Ethynylcyclopentanol


Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 156…159 °C
Melting Pt: 23…27 °C
Density: 0.962
Flash Pt: 48 °C(118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003858
CAS Number: 17356-19-3
EINECS: 241-385-7
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty