Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-p-toluate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-Toluic acid methyl ester4-Methylbenzoic acid methyl ester4-Toluic acid methyl esterMethyl-4-methylbenzoateMethyl 4-methylbenzoate

B23111-36 B23111-22 B23111-36 B23111-22
AAB23111-22EA 37.19 USD InStock
CAAAB23111-22 CAAAB23111-36 AAB23111-22 AAB23111-36
Methyl-p-toluate 99%
Methyl p-toluate


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 221…224 °C
Melting Pt: 32…35 °C
Density: 1.060
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008441
CAS Number: 99-75-2
EINECS: 202-784-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty