Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-5-nonanol 98%

Supplier: Thermo Scientific Chemicals

B23167-09 B23167-18 B23167-30 B23167-09 B23167-18 B23167-30
AAB23167-18EA 226.99 USD InStock
AAB23167-18 AAB23167-09 CAAAB23167-09 CAAAB23167-30 CAAAB23167-18 AAB23167-30
4-Methyl-5-nonanol 98%
4-Methyl-5-nonanol


Formula: C₁₀H₂₂O
MW: 158.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046696
CAS Number: 154170-44-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty