Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Propiolactone 97%

Supplier: Thermo Scientific Chemicals
Danger

B23197-06 B23197-03 B23197-06 B23197-14 B23197-14 B23197-03
AAB23197-06EA 267.05 USD InStock
CAAAB23197-14 CAAAB23197-03 AAB23197-06 AAB23197-03 AAB23197-14 CAAAB23197-06
β-Propiolactone 97%
β-Propiolactone


Formula: C₃H₄O₂
Boiling Pt: 161…163 °C
Melting Pt: –34 °C
Density: 1.146
Flash Pt: 70 °C(158 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005169
CAS Number: 57-57-8
EINECS: 200-340-1
UN: 3382
ADR: "6.1",I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty