Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylsulfonyl)aniline hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

B23202-22 B23202-14 B23202-22 B23202-06 B23202-14 B23202-06
AAB23202-06EA 65.59 USD InStock
AAB23202-06 CAAAB23202-22 CAAAB23202-14 CAAAB23202-06 AAB23202-14 AAB23202-22
4-(Methylsulfonyl)aniline hydrochloride 99%
4-(Methylsulphonyl)aniline hydrochloride


Formula: C₇H₁₀ClNO₂S
Melting Pt: 237…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00216490
CAS Number: 177662-76-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty