Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Phenylbutyric acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Phenylbutanoic acid(±)-2-Phenylbutanoic acid

B23222-22 B23222-22 B23222-36 B23222-36
AAB23222-22EA 61.69 USD InStock
AAB23222-22 CAAAB23222-22 CAAAB23222-36 AAB23222-36
(±)-2-Phenylbutyric acid 98%
(±)-2-Phenylbutyric acid


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 270…272 °C
Melting Pt: 40…44 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002667
CAS Number: 90-27-7
EINECS: 201-982-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty