Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminosalicylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Amino-2-hydroxybenzoic acid

B23289-22 B23289-22
AAB23289-22EA 60.39 USD InStock
CAAAB23289-22 AAB23289-22
4-Aminosalicylic acid 98+%
4-Aminosalicylic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: ca 135…145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007789
CAS Number: 65-49-6
EINECS: 200-613-5
Merck Index: 13,00476
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty