Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Mercaptosuccinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Thiomalic acidDL-Mercaptosuccinic acid(±)-Mercapto-1,4-butanedioic acid

B23301-14 B23301-36 B23301-22 B23301-22 B23301-14 B23301-36
AAB23301-14EA 23.52 USD InStock
CAAAB23301-22 AAB23301-14 AAB23301-22 CAAAB23301-14 CAAAAB23301-36 AAAB23301-36
(±)-Mercaptosuccinic acid 98%
(±)-Mercaptosuccinic acid


Formula: C₄H₆O₄S
MW: 150.16 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4
Merck Index: 12,09479
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty