Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-2-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

B23382-22 B23382-36 B23382-22 B23382-36
AAB23382-22EA 34.89 USD InStock
CAAAB23382-36 CAAAB23382-22 AAB23382-22 AAB23382-36
4-Chloro-2-nitrotoluene 99%
4-Chloro-2-nitrotoluene


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 242 °C
Melting Pt: 34…38 °C
Flash Pt: 120 °C(248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007215
CAS Number: 89-59-8
EINECS: 201-921-2
UN: 3457
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty