Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Cyclopenten-1-one 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-CyclopentenoneCyclopenten-2-oneCyclopent-2-enone

B23408-06 B23408-14 B23408-06 B23408-14
AAB23408-06EA 75.57 USD InStock
AAB23408-06 CAAAB23408-06 AAB23408-14 CAAAB23408-14
2-Cyclopenten-1-one 98%
2-Cyclopenten-1-one


Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 150 °C
Density: 1.01
Flash Pt: 42 °C(108 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001401
CAS Number: 930-30-3
EINECS: 213-213-0
UN: 1224
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty