Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

B23426-06 B23426-06 B23426-22 B23426-14 B23426-22 B23426-14
AAB23426-14EA 160.99 USD InStock
AAB23426-14 AAB23426-22 CAAAB23426-14 AAB23426-06 CAAAB23426-22 CAAAB23426-06
3-Fluorobenzonitrile 98%
3-Fluorobenzonitrile


Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 183 °C
Melting Pt: –16 °C
Density: 1.140
Flash Pt: 67 °C(152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001797
CAS Number: 403-54-3
EINECS: 206-963-5
UN: 3276
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty