Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Norvaline 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(-)-NorvalineH-D-Nva-OHD(-)-Norvaline

B23444-06 B23444-03 B23444-14 B23444-14 B23444-03 B23444-06
AAB23444-06EA 77.89 USD InStock
AAB23444-06 AAB23444-03 AAB23444-14 CAAAB23444-14 CAAAB23444-03 CAAAB23444-06
D-Norvaline 99%
D-Norvaline


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008097
CAS Number: 2013-12-9
EINECS: 217-936-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty