Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl octanoate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Caprylic acid ethyl esterOctanoic acid ethyl esterEthyl caprylate

B23450-22 B23450-36 B23450-22 B23450-36
AAB23450-22EA 24.5 USD InStock
AAB23450-22 CAAAB23450-36 CAAAB23450-22 AAB23450-36
Ethyl octanoate 99%
Ethyl octanoate


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 206…208 °C
Melting Pt: –47 °C
Density: 0.872
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty