Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-nitrobenzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

B23451-14 B23451-22 B23451-14 B23451-22
AAB23451-14EA 32.29 USD InStock
AAB23451-14 AAB23451-22 CAAAB23451-14 CAAAB23451-22
4-Chloro-3-nitrobenzophenone 99%
4-Chloro-3-nitrobenzophenone


Formula: C₁₃H₈ClNO₃
MW: 261.66 g/mol
Boiling Pt: 233…235 °C/12 mm
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00007081
CAS Number: 56107-02-9
EINECS: 259-995-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty