Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethylhexanal 97%

Supplier: Thermo Scientific Chemicals
Warning

B23471-18 B23471-30 B23471-30 B23471-18
AAB23471-30EA 136.92 USD InStock
AAB23471-30 CAAAB23471-30 AAB23471-18 CAAAB23471-18
2-Ethylhexanal 97%
2-Ethylhexanal


Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 163 °C
Melting Pt: < –100 °C
Density: 0.819
Flash Pt: 50 °C(122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006987
CAS Number: 123-05-7
EINECS: 204-596-5
UN: 1191
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty