Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-glutamic acid 99%

Supplier: Thermo Scientific Chemicals

B23621-22 B23621-14 B23621-14 B23621-22
AAB23621-22EA 68.49 USD InStock
CAAAB23621-14 AAB23621-22 CAAAB23621-22 AAB23621-14
N-Acetyl-L-glutamic acid 99%
N-Acetyl-L-glutamic acid


Formula: C₇H₁₁NO₅
Melting Pt: 194…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00002802
CAS Number: 1188-37-0
EINECS: 214-708-4
Order Now
Specification Test Results
Optical rotation -16.5° (c=1 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty