Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) lactate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

B23643-22 B23643-14 B23643-36 B23643-36 B23643-14 B23643-22
AAB23643-14EA 29.89 USD InStock
AAB23643-14 AAB23643-36 CAAAB23643-36 CAAAB23643-14 AAB23643-22 CAAAB23643-22
Nickel(II) lactate tetrahydrate 98%
Nickel(II) lactate tetrahydrate


Formula: C₆H₁₀NiO₆·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD02683106
UN: 3077
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty