Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutaronitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

B23663-18 B23663-18
AAB23663-18EA 127.54 USD InStock
CAAAB23663-18 AAB23663-18
Glutaronitrile 99%
Glutaronitrile


Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 286…288 °C
Melting Pt: –29 °C
Density: 0.990
Flash Pt: 112 °C(234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001970
CAS Number: 544-13-8
EINECS: 208-861-6
UN: 3276
ADR: "6.1",III
Merck Index: 13,04487
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty